Specifications
- Antibody type:Primary
- Antigen symbol:NNP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8568
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Novel nuclear protein 1 (NNP-1), also known as RRP1-like protein or Nucleolar protein Nop52, is a 461 amino acid protein belonging to the RRP1 family. Localized to the nucleolus, NNP-1 has simian virus 40-type and bipartite nuclear localization signals and four coiled-coil domains within its C-terminal region. NNP-1 has been found to play an important role in the generation of 28S rRNA in the late processing steps of ribosome biogenesis. At the end of mitosis, nucleolar proteins assemble in a sequential order during the rebuilding of the nucleolus. NNP-1 assembles after Fibrillarin and C23, and simultaneously with B23 and POP1 in the prenucleolar body pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NNP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat