Anti-S100 A7A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-6575R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium binding Protein A7A
- Antigen symbol:S100 A7A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:381493
- Antigen synonyms:Gm1020|S100A7L1|S100A7f|S100a15|S100a17l1|AY465109|S100a15a
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:40-80/108
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Fetal ear, skin, and tongue and human cell lines. Highly up-regulated in psoriatic epidermis. Also highly expressed in the urine of bladder squamous cell carcinoma (SCC) bearing patients.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: S100 A7A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat