Anti-APOBR Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-6336R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein B Receptor
- Antigen symbol:APOBR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:55911
- Antigen synonyms:APOB48R|APOB100R
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
APOB48R is a macrophage receptor that binds to the apolipoprotein B48 of dietary triglyceride (TG) rich lipoproteins. It may provide essential lipids, lipid soluble vitamins and other nutrients to reticuloendothelial cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOBR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat