Anti-CASC5 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-5959R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cancer Susceptibility Candidate 5
- Antigen symbol:CASC5
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:57082
- Antigen synonyms:D40|PPP1R55|Spc7|CT29|hKNL-1|KNL1|AF15Q14|hSpc105
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Part of the MIS12 complex, which may be fundamental for kinetochore formation and proper chromosome segregation during mitosis. Four known isoforms exist.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CASC5
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat