Anti-TBC1D4 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-5893R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TBC1 Domain Family Member 4
- Antigen symbol:TBC1D4
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:9882
- Antigen synonyms:CDC16) domain-containing protein|Acrg embryonic lethality minimal region ortholog|AS160|TBC1 domain family member 4|akt substrate of 160 kDaStore at 4║C for 1-2 weeks. For long-term storage|store at -20║C.|TBC (Tre-2|RP11-159J2.3|DKFZp779C0666|BUB2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May act as a GTPase-activating protein for RAB2A, RAB8A, RAB10 and RAB14. Isoform 2 promotes insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane, thus increasing glucose uptake.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TBC1D4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat