Anti-ADAM19 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5850R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase Domain 19
- Antigen symbol:ADAM19
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:8728
- Antigen synonyms:MLTNB|FKSG34|Disintegrin and metalloproteinase domain-containing protein 19|Meltrin-beta|ADAM19|Metalloprotease and disintegrin dendritic antigen marker|MADDAM|ADAM 19
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Participates in the proteolytic processing of beta-type neuregulin isoforms which are involved in neurogenesis and synaptogenesis, suggesting a regulatory role in glial cell. Also cleaves alpha-2 macroglobulin. May be involved in osteoblast differentiation and/or osteoblast activity in bone (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADAM19
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat