Anti-LSP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-5156R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Lymphocyte-specific Protein 1
- Antigen symbol:LSP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:4046
- Antigen synonyms:WP34|pp52|F-actin binding and cytoskeleton associated protein|leukocyte-specific protein 1|lymphocyte-specific protein pp52|52 kDa phosphoprotein|leufactin (leukocyte F-actin binding protein)|lymphocyte-specific protein 1|LSP1|47 kDa actin binding protein|lymphocyte-specific antigen WP34
- Modification:phospho-Ser252
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May play a role in mediating neutrophil activation and chemotaxis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LSP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human