Anti-WFDC2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-4626R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:WAP four-disulfide core Domain 2
- Antigen symbol:WFDC2
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:67701
- Antigen synonyms:epididymal protein 4|WFDC2|four-disulfide core|EDDM4|WAP domain containing protein HE4-V4|WAP four-disulfide core domain 2|WAP5|MGC57529|epididymal secretory protein E4|epididymis-specific|major epididy|whey-acidic protein type|dJ461P17.6|HE4
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:110-160/174
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Broad range protease inhibitor.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: WFDC2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat