Anti-KLK11 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-3799R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 11
- Antigen symbol:KLK11
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:11012
- Antigen synonyms:Kallikrein-11 inactive chain 2|Serine protease 20|KLK11|Kallikrein-11 inactive chain 1|PRSS20|Kallikrein11|Hippostasin|UNQ649/PRO1279|Kallikrein 11|TLSP|hK11|Trypsin-like protease|Kallikrein-11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Kallikrein 11 has two alternative splicing isoforms, known as the brain type and prostate type. Elevated serum levels of hK11 have been found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of this biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat