Anti-PDP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-3966R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Pyruvate Dehyrogenase Phosphatase Catalytic Subunit 1
- Antigen symbol:PDP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:54704
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
PPM2C is a protein similar to members of the protein phosphatase 2C (PP2C) family. It is a catalytic subunit of the mitochondrial matrix phosphatase and is involved in reactivation of the pyruvate dehydrogenase complex.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat