Specifications
- Antibody type:Primary
- Antigen name:Rotavirus protein NSP3
- Antigen symbol:NSP3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:SH2D3C|Cas/HEF1 associated signal transducer|FLJ39664|SH2D3_HUMAN|SH2 domain containing protein 3C|CHAT|SH2 domain containing Eph receptor binding protein 1|Novel SH2-containing protein 3|SH2 domain containing 3C|PRO34088|Novel SH2 containing protein 3|SH2 domain-containing protein 3C|Shep1.|NSP 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Tumor necrosis factor (TNF) is a pleiotropic cytokine whose function is mediated through two distinct cell surface receptors (1,2). These receptors, designated TNF-R1 and TNF-R2, are expressed on most cell types (1,3). The majority of TNF functions are primarily mediated through TNF-R1 (1,4). FAN (for factor associated with neutral sphingomyelinase (N-SMase) activation) is an intermediate protein that interacts with TNF-R1 to initiate TNF signaling events. FAN binds to TNF-R1 at the cytoplasmic NSD (N-SMase activating domain), which results in the initiation of the N-SMase pathway (5). N-SMase has been shown to be involved in TNF-induced Raf-1 activation (6). FAN contains four carboxy-terminal WD-repeat domains which appear to be involved in protein-protein interaction. The FAN WD-repeats may mediate the interaction between FAN and TNF-R1 (5).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NSP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat