Order Entry
Ireland
ContactUsLinkComponent
Anti-GOLGA4 Rabbit Polyclonal Antibody (Cy5®)
Anti-GOLGA4 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13487R-CY5
Supplier:  Bioss
Anti-GOLGA4 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13487R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Golgin Subfamily A Member 4
  • Antigen symbol:
    GOLGA4
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2803
  • Antigen synonyms:
    p230|GCP2|MU-RMS-40.18|CRPF46|GOLG
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Golgin 245 is also known as p230, GCP2, GOLG or golgi autoantigen and is a 2,230 amino acid protein that is expressed as three isoforms. Golgin 245 is localized to the cytoplasm in cells and is a member of the golgin family, all of which are peripheral membrane proteins associated with the Golgi complex. Golgin 245 has a carboxyl-terminal GRIP domain, which attaches to the trans-Golgi network (TGN) and TGN-derived vesicles. It is thought that the interaction that takes place between golgin 245 and MACF1, which cross-links microtubules to the Actin cytoskeleton, allows proteins to be transported from the TGN to the cell periphery. against golgin 245 are associated with two chronic disorders known as Sjé°ƒren's syndrome and Hepatitis B.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Golgin subfamily A member 4/Golgin 245
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat