Specifications
- Antibody type:Primary
- Antigen name:Elongation factor 1-delta
- Antigen symbol:EEF1D
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:1936
- Antigen synonyms:FP1047|EF-1D|EF1D
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a subunit of the elongation factor-1complex, which is responsible for the enzymatic delivery ofaminoacyl tRNAs to the ribosome. This subunit, delta, functions asguanine nucleotide exchange factor. It is reported that followingHIV-1 infection, this subunit interacts with HIV-1 Tat. Thisinteraction results in repression of translation of host cellproteins and enhanced translation of viral proteins. Severalalternatively spliced transcript variants encoding multipleisoforms have been found for this gene. Related pseudogenes havebeen defined on chromosomes 1, 6, 7, 9, 11, 13, 17, 19.[provided byRefSeq, Aug 2010].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EEF1D
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat