Order Entry
Ireland
ContactUsLinkComponent
Anti-PC2 Rabbit Polyclonal Antibody (Cy7®)
Anti-PC2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-11533R-CY7
Supplier:  Bioss
Anti-PC2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-11533R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proprotein Convertase 2
  • Antigen symbol:
    PC2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5126
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    255-290/638
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Responsible for the release of glucagon from proglucagon in pancreatic A cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PC2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat