Anti-ARC Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-0385R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Activity-regulated Cytoskeleton-associated Protein
- Antigen symbol:ARC
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:23237
- Antigen synonyms:Nop30|Myp|Muscle-enriched cytoplasmic protein|Apoptosis Repressor with CARD|Nucleolar protein of 30 kDa|Arg3.1|Nucleolar protein 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:100-150/396
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Required for consolidation of synaptic plasticity as well as formation of long-term memory. Regulates endocytosis of AMPA receptors in response to synaptic activity. Required for homeostatic synaptic scaling of AMPA receptors (By similarity). Plays a role in the regulation of cell morphology and cytoskeletal organization. Required in the stress fiber dynamics and cell migration.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARC
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat