Long flange with weld stub
EDWAC10514316
- Description:Long flange fitting, NW25, 70 mm, operating pressure range 10-7 mbar to 17 bar
- Environmentally Preferable:
- Pk:1
This flange fittings made from stainless steel 316L are ideal for rugged corrosion resistance in semiconductor processing and offer excellent repeatability in high vacuum applications.
- Quality control of elastomer specifications
- Fit-and-forget dependability