Anti-CHD3 Mouse Monoclonal Antibody
BWRLMB0061
- Antibody type:Primary
- Antigen name:Chromodomain helicase DNA binding protein 3
- Antigen symbol:CHD3
- Clonality:Monoclonal
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the C-terminus of Human CHD3.
This antibody detects endogenous levels of CHD3 (C-terminus) and does not cross-react with related proteins
Type: Primary
Antigen: CHD3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human