Anti-PLA2G1B Rabbit Polyclonal Antibody
Catalog # BWRLBS7786
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PLA2G1B
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human PLA2G1B.
PLA2G1B pAb detects endogenous levels of PLA2G1B protein.
Type: Primary
Antigen: PLA2G1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat