Anti-Cerebellin 4 Rabbit Polyclonal Antibody [clone: K178]
Catalog # BWRLBS3282
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Cerebellin 4
- Clonality:Polyclonal
- Clone:K178
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 148-197 of Human Cerebellin 4.
Cerebellin 4 (K178) pAb detects endogenous levels of Cerebellin 4 protein.
Type: Primary
Antigen: Cerebellin 4
Clonality: Polyclonal
Clone: K178
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat