Anti-MCH-2R Rabbit Polyclonal Antibody [clone: M332]
Catalog # BWRLBS2629
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MCH-2R
- Clonality:Polyclonal
- Clone:M332
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 300-348 of Human MCH-2R.
MCH-2R (M332) pAb detects endogenous levels of MCH-2R protein.
Type: Primary
Antigen: MCH-2R
Clonality: Polyclonal
Clone: M332
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human