Anti-TCR beta Rabbit Polyclonal Antibody [clone: K115]
BWRLBS2943
- Antibody type:Primary
- Antigen symbol:TCR beta
- Clonality:Polyclonal
- Clone:K115
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 81-130 of Human TCR beta.
TCR ? (K115) pAb detects endogenous levels of TCR ? protein.
Type: Primary
Antigen: TCR beta
Clonality: Polyclonal
Clone: K115
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human