Piperacillin sodium salt, white powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CAS Number:59703-84-3
:MFCD00917471
Formula:C₂₃H₂₇N₅O₇S
MDL Number:MFCD00056867
Molecular Weight:539.54 g/mol
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
Specification Test Results
Frequently Bought Together