Phytohemagglutinin (from Red kidney bean), MP Biomedicals
Catalog # ICNA08791501
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 mg
- Source:Red kidney bean
- Species:
- Environmentally Preferable:
- Protein/peptide name:Phytohemagglutinin
- Formulation:Lyophilised powder
Specifications
About this item
Phytohemagglutinin PHA agglutinates the erythrocytes of all human blood groups as well as those of rabbit, dog, cat, mouse, sheep, and guinea pig.