Order Entry
Ireland
ContactUsLinkComponent
RNA ladder, high range, RiboRuler™
RNA ladder, high range, RiboRuler™
  SM1821
 :  
undefined
RNA ladder, high range, RiboRuler™
  SM1821
 :  
  • Pk:
    1 KIT
  • Description:
    RiboRuler™ High Range RNA Ladder, 5×20 µl
  • Environmentally Preferable:
RiboRuler™ high range RNA ladder allows precise sizing of RNA molecules in denaturing agarose gels and Northern blotting.
  • Easy-to-remember band sizes and quantities
  • Sharp bands of uniform intensity
  • Predetermined concentration of ladder bands allows for approximate RNA quantification
  • Available in both conventional and ready-to-use format
RiboRuler™ high range RNA ladder is composed of chromatography-purified transcripts that makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically. To minimise the possibility of contamination during the use, the ladder is aliquotted in smaller volumes.
 : Supplied with 2X RNA loading dye.