Alcalase (from Bacillus licheniformis)
126741-500
:
- Pk:500 mL
- Source:Bacillus licheniformis
- Species:
- Environmentally Preferable:
- Storage conditions:2...8 °C
- Form:Dark brown
- Enzyme name:Alcalase
- Enzyme activity:≥0,75 Anson u/ml
- pH:7,5
A serine endopeptidase that consists primarily of subtilisin A. Suitable for the hydrolysis of proteins.
One unit is defined as the amount of enzyme that will release 1,0 µmol L-tyrosine from hemoglobin per min at 25 °C.