Epidural anaesthesia needle, Perican®
Catalog # BRAU4512383
Supplier: B. Braun
Specifications
- Gauge:G 18
- Colour:Pink
- Description:Epidural anaesthesia needle with Tuohy bevel
- Length:83 mm
- Environmentally Preferable:
- Pk:100
Specifications
About this item
Epidural anaesthesia needle with Tuohy-type bevel.
- Design for different tissue layers
- Control of needle advancement and small wings
- Transparent needle with colour-coded stylet hub
Suitable for all procedures specially for epidural anesthesia.