Order Entry
Ireland
ContactUsLinkComponent
Anti-HIF-PH3 Mouse Monoclonal Antibody [clone: EG188e/d5]
Anti-HIF-PH3 Mouse Monoclonal Antibody [clone: EG188e/d5]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-HIF-PH3 Mouse Monoclonal Antibody [clone: EG188e/d5]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-30440SS
  • NOVUNBP1-30440
  • Antigen symbol:
  • HIF-PH3
  • HIF-PH3
  • Antigen name:
  • HIF Prolyl Hydroxylase 3
  • HIF Prolyl Hydroxylase 3
  • Antigen synonyms:
  • egl nine homolog 3|prolyl hydroxylase domain-containing protein 3|HIF-PH3|egl nine-like protein 3 isoform|HIF-prolyl hydroxylase 3|HIF prolyl hydroxylase 3|PHD3|HPH-1|HIFPH3|HPH-3|hypoxia-inducible factor prolyl hydroxylase 3
  • egl nine homolog 3|prolyl hydroxylase domain-containing protein 3|HIF-PH3|egl nine-like protein 3 isoform|HIF-prolyl hydroxylase 3|HIF prolyl hydroxylase 3|PHD3|HPH-1|HIFPH3|HPH-3|hypoxia-inducible factor prolyl hydroxylase 3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • EG188e/d5
  • EG188e/d5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 112399
  • 112399
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together