Anti-SLCO3A1 Rabbit Polyclonal Antibody
BOSSBS-11802R
: Bioss
- Antibody type:Primary
- Antigen name:Solute Carrier Organic Anion Transporter Family member 3A1
- Antigen symbol:Solute carrier organic anion transporter family, member 3A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Solute carrier organic anion transporter family member 3A1|SO3A1_HUMAN.|FLJ40478|SLC21A11|OATP D|OATP3A1|OATPD|Organic anion transporter polypeptide related protein 3|OATP RP3|Organic anion transporting polypeptide D|Solute carrier family 21 member 11|Sodium independent organic anion transporter D|OATPRP3|PGE1 transporter|Solute carrier family 21 organic anion transporter member 11
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Type: Primary
Antigen: SLCO3A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat