Anti-GroEL Mouse Monoclonal Antibody [clone: 9A1/2]
New Product
GroEL and GroES are E.coli chaperonins, which are homologs of mitochondrial chaperonins Hsp60 and Hsp10. GroEL is a double toriodal assembly of 14 identical subunits which form two heptameric rings stacked back-to-back, with a cavity at each end. GroEL and its co-chaperonin GroES facilitate protein folding with an ATP-dependent mechanism.
- Dilution range: Western blot (1:10,000, ECL)
- Detects a band of ~60kDa by western blot
Type: Primary
Antigen: GroEL
Clonality: Monoclonal
Clone: 9A1/2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: E.coli
- Catalog No:
- ENZOADISPS870F
- ENZOADISPS870D
- Antigen symbol:
- HSPD1
- HSPD1
- Antigen name:
- Heat shock protein family D (Hsp60) member 1
- Heat shock protein family D (Hsp60) member 1
- Antigen synonyms:
- GROEL|HSP65|HLD4|HSP-60|HuCHA60|HSP60|CPN60|SPG13
- GROEL|HSP65|HLD4|HSP-60|HuCHA60|HSP60|CPN60|SPG13
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 9A1/2
- 9A1/2
- Reactivity:
- Bacteria
- Bacteria
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- E. coli GroEL.
- E. coli GroEL.
- Form:
- Liquid
- Liquid
- Purification:
- purified by Protein G affinity chromatography
- purified by Protein G affinity chromatography
- Storage buffer:
- PBS containing 50% glycerol and 0,09% sodium azide
- PBS containing 50% glycerol and 0,09% sodium azide
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together