Glycerine solution 80%, sterile
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
Boiling Pt:182 °C (20 mmHg)) (lit.)
Melting Pt:18.6
Density:1.25 g/mL (lit.)
Molecular Weight:92.09 g/mol
Flash Pt:177
Merck Index:13,04497
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Heavy Metals:≤2 ppm
- Water Content:≤0,5%
- Storage Conditions:Store at Room Temperature (15-30 °C)
- Assay:≥99,5% (GC), corrected for water
- Concentration:0,8
- Auto ignition:698 °F (Lit.)
Specification Test Results
Frequently Bought Together