Fuchsin (acid), powder C.I. 42685
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: Rubin S, CN DcR-8, Acid Violet 19, Acid Fuchsin, Acid Fuchsin sodium salt, Fuchsin S, CI 42685, Acid Magenta, 3-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acid, Benzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)
CAS Number:3244-88-0
:
Formula:C₂₀H₁₇N₃Na₂O₉S₃
MDL Number:MFCD00013286
Molecular Weight:585.55 g/mol
Merck Index:12,00109
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
- UV/VIS: l max (water):543 - 547 nm
Specification Test Results
Frequently Bought Together