Reversed phase HPLC columns, Synergi™ Hydro-RP
Synergi™ Hydro-RP columns have polar embedded C18 phase that provides extreme hydrophobic and slight polar selectivity.
- Ideal for separation of weakly hydrophobic compounds
- Stable in 100% aqueous mobile phase
Frequently Bought Together