Antibiotic assay discs, multodisc
Multodisc with eight antimicrobial agents on a ring.
- Each specially for a microorganism
Amikacin (AK), Ampicillin (AMP), Ampicillin-Sulbactam (AMS), Carbenicillin (CAR), Cefoxitin (FOX), Ceftazidime (CAZ), Cefuroxime (CXM), Chloramphenicol (C), Clindamycin (CD), Erythromycin (E), Gentamicin (CN), Levofloxacin (LEV), Linezolid (LNZ), Metronidazole (MTZ), Netilmicin (NET), Nitrofurantoin (F), Norfloxacin (NOR), Penicillin G (P), Piperacillin-Tazobactam (TZP), Rifampicin (RD), Streptomycin (S), Teicoplanin (TEC), Tetracycline (TE), Ticarcillin (TC), Tobramycin (TOB), Trimethoprim-Sulfamethoxazole (SXT), Vancomycin (VA). The numbers shows the concentration in μg.
- Environmentally Preferable:
Frequently Bought Together