4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) mixture of isomers 99% (mixture of isomers)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Bis(4-amino-3-methylcyclohexyl)methane, 4,4'-Methylenebis(2-methylcyclohexylamine), 4,4'-Methylenebis(1-amino-2-methylcyclohexane), 4,4'-Diamino-3,3'-dimethyldicyclohexylmethane
CAS Number:6864-37-5
:
Formula:C₁₅H₃₀N₂
MDL Number:MFCD00075535
Boiling Pt:93…100 °C (760 mmHg)
Density:0.94 g/cm³
Molecular Weight:238.42 g/mol
Flash Pt:113 °C
UN:2927
ADR:6.1 II
:GHS05
:GHS06
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4980 to 1.5000 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together