Potassium (cyanomethyl)trifluoroborate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Potassium (cyanomethyl)(trifluoro)borate(1-), potassium (cyanomethyl)trifluoroboranuide
CAS Number:888711-58-8
:
Formula:C₂H₂BF₃KN
MDL Number:MFCD11052818
Molecular Weight:146.949 g/mol
Melting Pt:151…156 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Solid
- Infrared spectrum:Conforms
- Melting point:144 to 156 °C
- NMR (1H-NMR):Consistent with structure
Specification Test Results
Frequently Bought Together