Methanol, Technical Grade
Catalog # ACRO444310050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.3280 to 1.3300 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥99 %
- Ethanol:≤0.5 %
Specification Test Results
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
:
:32.04 g/mol
UN:1230
:GHS06
:GHS08
:GHS02
Melting Pt:–98 °C
Storage Temperature:Ambient
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595
Boiling Pt:64.7 °C (760 mmHg)
Density:0.791 g/cm³
Flash Pt:12 °C
ADR:3 II
Merck Index:13,05984