4-iodobutanenitrile 97%
ACRO390390010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5350 to 1.5380 (20°C, 589 nm)
- GC:≥96.0 %
: 4-Iodobutyronitrile, 4-Iodobutanenitrile
:
:195.003 g/mol
:3276
:GHS07
:Ambient
:6727-73-7
:C₄H₆IN
:MFCD00039501
:1.826 g/cm³
:6.1 III