1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: SIPr.HCl, 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium chloride, 1,3-Bis(2,6-diisopropylphenyl)imidazolidinium chloride
CAS Number:258278-25-0
:
Formula:C₂₇H₃₉ClN₂
MDL Number:MFCD07369796
Molecular Weight:427.07 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥88 %
Specification Test Results
Frequently Bought Together