Order Entry
Ireland
ContactUsLinkComponent
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 97%
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 97%
Certificates
undefined
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 97%
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White to almost white Powder and/or chunks
  • Infrared spectrum:
    Conforms
  • Water:
    ≤17 % (K.F.)
  • Titration with HClO4:
    ≥96.0 % (on dry substance)