Order Entry
Ireland
ContactUsLinkComponent
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
Certificates
undefined
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to light beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    117 to 121 °C
  • Specific optical rotation:
    +41.5° to +47.5° (20 °C, 589 nm) (c=1, CH3OH)
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together