Acetaldehyde ammonia trimer 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aldehyde Ammonia Trimer, 2,4,6-Trimethylhexahydro-1,3,5-triazine Trihydrate, Hexahydro-2,4,6-trimethyl-1,3,5-triazine Trihydrate, 2,4,6-trimethyl-1,3,5-triazinane trihydrate
CAS Number:58052-80-5
:
Formula:C₆H₂₁N₃O₃
MDL Number:MFCD00149559
Boiling Pt:110 °C (760 mmHg)
Molecular Weight:183.25 g/mol
Melting Pt:95…97 °C
Storage Temperature:Ambient
UN:1841
ADR:9 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light beige Crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together