4-Piperidone monohydrate hydrochloride 98%
: 4,4-Piperidinediol Hydrochloride, 4,4-Dihydroxypiperidine Hydrochloride, 4-Ketopiperidine Monohydrate Hydrochloride, 4-Piperidinone monohydrate hydrochloride, Piperidin-4-one hydrate hydrochloride, 4-Azacyclohexanone Monohydrate Hydrochloride
:40064-34-4
:C₅H₉NO·xHCl·yH₂O
:MFCD00012776
:99.13 g/mol (free base)
:89…99 °C
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:Off-white to yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:89 to 99 °C
- Water:10.5 to 13 %
- TLC:≥97.5 %
- Titration with HClO4:≥97.5 % (On anhydrous substance)
Frequently Bought Together