Order Entry
Ireland
ContactUsLinkComponent
Mitomycin C (2 mg/48 mg NaCl)
Mitomycin C (2 mg/48 mg NaCl)
Catalog # ACRO226940020
CAS Number:  
undefined
Mitomycin C (2 mg/48 mg NaCl)
Catalog # ACRO226940020
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    2 mg
  • Seq:
    2145
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Blue to violet Powder
  • HPLC:
    ≥99 % with regard to the non-formulated material

Specification Test Results

About this item

Synonyms: Mitomycin, Ametycin
 :
 :334.33 g/mol
UN:2811
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:50-07-7
Formula:C₁₅H₁₈N₄O₅
MDL Number:MFCD00078109
Melting Pt:>360 °C
ADR:6.1 III