Test kits, photometric, COD, LCK cuvette tests
: Hach
Cuvette tests for water testing applications. Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient, error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour coding
Storage conditions: 15 to 25 °C (protect from light)
: ISO 6060-1989, DIN 38409-H41-H44
: Cuvette tests are officially approved for many legally required consent limits and with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.
- Environmentally Preferable:
COD cuvette tests for salt water
Frequently Bought Together