VWR®, Absorbents - Mats, Rolls and Socks, for Chemical Based Spills
VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
- Absorbs a wide range of chemicals, including acids and caustics
- Chemically inert: Product does not degrade or cause a dangerous chemical reaction with the adsorbed liquid
- Green colour alerts that product is being used for hazardous liquids
- Green colour allows user to identify and separate hazardous waste for disposal, resulting in lower disposal costs
- Cannot be left outdoors as they will absorb water
For use with chemicals.
Colour: Green.
Frequently Bought Together