Anti-MYCN Mouse Monoclonal Antibody [clone: NCM-II 100]
Catalog # PIEAMA1170
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:V-Myc Avian Myelocytomatosis Viral Oncogene Neuroblastoma Derived Homolog
- Antigen symbol:n-Myc
- Clonality:Monoclonal
- Clone:NCM-II 100
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Purified antibody
- Form:Liquid
- Gene ID:4613
- Antigen synonyms:Class E Basic Helix-Loop-Helix Protein|Oncogene NMYC|N-Myc Proto-Oncogene Protein|bHLHe37|NMYC proto oncogene protein|Neuroblastoma MYC Oncogene|MYCN
- Storage buffer:PBS, 30% glycerol, 1mg/ml BSA, 0.05% sodium azide, pH 7.2-7.4
- Molecular weight:49.5 kDa
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Immunogen:human fusion protein
- Purification:Protein A
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen Symbol: MYCN
Antigen Name: v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
Clonality: Monoclonal
Clone: NCM-II 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human