2-(Trifluoromethyl)pyridin-4-ol 97%
Catalog # H64380.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Hydroxy-2-(trifluoromethyl)pyridine
CAS Number:170886-13-2
:
Formula:C₆H₄F₃NO
:163.10 g/mol
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD01571202