3-Bromofuro[3,2-b]pyridine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92404-63-2
:
Formula:C₇H₄BrNO
MDL Number:MFCD19687763
Molecular Weight:198.02 g/mol
Melting Pt:51…53 °C
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together