Amberlite® IRA-96 ion-exchange resin
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39409-19-3
:
MDL Number:MFCD00166430
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together