1-Hexylboronic acid pinacol ester 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS Number:86308-26-1
:
Formula:C₁₂H₂₅BO₂
MDL Number:MFCD08457665
Density:1.167
Molecular Weight:212.14 g/mol
Flash Pt:84 °C (183 °F)
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together